Assay Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025911.2 | NP_080187.2 | 6 - 7 | 780 | 77 | |
| GenBank mRNA | AK007917.1 | - | 6 - 7 | 781 | 77 | |
| BC006734.1 | - | 6 - 7 | 685 | 3156328 | 77 | |
| BC024946.1 | - | 1 - 2 | 38 | 4010938 | 77 | |
| BC049073.1 | - | 6 - 7 | 749 | 5707404 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024946.1 transcripts.
Set Membership: |
|

Ccdc91
Genomic Map