Assay Details
| Gene Symbol: Agbl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048189.4 | NP_001041654.2 | 10 - 11 | 1278 | 72 | |
| NM_001284190.1 | NP_001271119.1 | 10 - 11 | 1278 | 72 | ||
| NM_030231.3 | NP_084507.1 | 10 - 11 | 1278 | 72 | ||
| GenBank mRNA | DQ867039.1 | - | 10 - 11 | 1185 | 72 | |
| DQ867040.1 | - | 10 - 11 | 1185 | 72 | ||
| FN429928.1 | - | 10 - 11 | 1104 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agbl4
Genomic Map