Assay Details
| Gene Symbol: Ppp1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163103.1 | NP_001156575.1 | 9 - 10 | 891 | 78 | |
| GenBank mRNA | AK133227.1 | - | 8 - 9 | 2186 | 78 | |
More Information
Set Membership: |
|

Ppp1r36
Genomic Map