Assay Details
| Gene Symbol: Ppp1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163103.1 | NP_001156575.1 | 10 - 11 | 1083 | 100 | |
| GenBank mRNA | AK133227.1 | - | 9 - 10 | 2378 | 100 | |
| AV274491.2 | - | 2 - 3 | 331 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521564 | NM_001163103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521564 | AK133227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521564 | NM_001163103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521564 | AK133227.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759808 | AK033518.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759808 | NM_178189.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | AK136173.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | AK145374.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | AK155990.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | AK158771.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | AK159081.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | BC030188.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | NM_001024806.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | U19891.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509058 | U19892.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92407 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99568 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12988 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165189 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521564 | NM_001163103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521564 | AK133227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521564 | NM_001163103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521564 | AK133227.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165189 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07671355 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07671339 | - | - |
| Whole Mouse Genome G4122A | A_52_P521564 | NM_001163103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521564 | AK133227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521564 | NM_001163103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521564 | AK133227.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09907544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10161883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09937256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09933235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10080788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09818918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09770767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09937259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09970479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553914 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1456823_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10396656 | - | - |
| Mouse Genome 430 2.0 Array | 1456823_at | - | - |
| Mouse Genome 430 2.0 Array | 1456823_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210762_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210762_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200004837.mm.1 | NM_001163103.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004837.mm.1 | NM_001163103.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004837.mm.1 | TC1200000680.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200004836.mm.1 | NM_001163103.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004836.mm.1 | NM_001163103.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004836.mm.1 | TC1200000680.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456823_PM_at | - | - |
| Mouse Expression Set 430 | 1456823_at | - | - |
| Murine Genome U74v2 Set | 167547_PM_at | - | - |
| Murine Genome U74v2 Set | 167547_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456823_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456823_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 629101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221094 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2526407 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2526571 | - | - |

Ppp1r36
Genomic Map