Assay Details
| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035231.1 | - | 1 - 2 | 135 | 88 | |
| BB637493.1 | - | 1 - 2 | 430 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035231.1 transcripts.
Set Membership: |
|

Etv6
Genomic Map