Assay Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015099.2 | NP_001015099.2 | 13 - 14 | 1880 | 136 | |
| NM_001167963.1 | NP_001161435.1 | 14 - 15 | 1949 | 136 | ||
| NM_001167964.1 | NP_001161436.1 | 12 - 13 | 1598 | 136 | ||
| GenBank mRNA | AK031333.1 | - | 12 - 13 | 1458 | 136 | |
| AK129332.1 | - | 13 - 14 | 1783 | 136 | ||
| BC086455.1 | - | 13 - 14 | 1753 | 30458160 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

G2e3
Genomic Map