Assay Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015099.2 | NP_001015099.2 | 8 - 9 | 971 | 91 | |
| NM_001167963.1 | NP_001161435.1 | 8 - 9 | 971 | 91 | ||
| NM_001167964.1 | NP_001161436.1 | 8 - 9 | 971 | 91 | ||
| GenBank mRNA | AK031333.1 | - | 8 - 9 | 831 | 91 | |
| AK033513.1 | - | 8 - 9 | 913 | 91 | ||
| AK084256.1 | - | 8 - 9 | 842 | 91 | ||
| AK129332.1 | - | 8 - 9 | 874 | 91 | ||
| BC086455.1 | - | 8 - 9 | 844 | 30458160 | 91 | |
More Information
Set Membership: |
|

G2e3
Genomic Map