Assay Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015099.2 | NP_001015099.2 | 9 - 10 | 1096 | 77 | |
| NM_001167963.1 | NP_001161435.1 | 9 - 10 | 1096 | 77 | ||
| NM_001167964.1 | NP_001161436.1 | 9 - 10 | 1096 | 77 | ||
| GenBank mRNA | AK031333.1 | - | 9 - 10 | 956 | 77 | |
| AK033513.1 | - | 9 - 10 | 1038 | 77 | ||
| AK084256.1 | - | 9 - 10 | 967 | 77 | ||
| AK129332.1 | - | 9 - 10 | 999 | 77 | ||
| BC086455.1 | - | 9 - 10 | 969 | 30458160 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

G2e3
Genomic Map