Assay Details
| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013845.4 | NP_038873.2 | 1 - 2 | 514 | 137 | |
| GenBank mRNA | AK046699.1 | - | 1 - 2 | 376 | 137 | |
| AK049369.1 | - | 1 - 2 | 515 | 137 | ||
| AK080896.1 | - | 1 - 2 | 462 | 137 | ||
| AK087548.1 | - | 1 - 2 | 137 | |||
| BC095933.1 | - | 1 - 2 | 463 | 137 | ||
| BC138794.1 | - | 1 - 2 | 499 | 8861816 | 137 | |
| BC138810.1 | - | 1 - 2 | 499 | 8861832 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046699.1, AK049369.1, AK080896.1, BC095933.1, BC138794.1, BC138810.1, NM_013845.4 transcripts.
Set Membership: |
|

Ror1
Genomic Map