Assay Details
| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013845.4 | NP_038873.2 | 6 - 7 | 1361 | 66 | |
| GenBank mRNA | AB010383.1 | - | 6 - 7 | 933 | 66 | |
| AK046699.1 | - | 6 - 7 | 1223 | 66 | ||
| AK049369.1 | - | 6 - 7 | 1362 | 66 | ||
| AK080896.1 | - | 6 - 7 | 1309 | 66 | ||
| AK087548.1 | - | 6 - 7 | 1294 | 66 | ||
| BC138794.1 | - | 6 - 7 | 1346 | 8861816 | 66 | |
| BC138810.1 | - | 6 - 7 | 1346 | 8861832 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ror1
Genomic Map