Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198631.2 | NP_941033.2 | 4 - 5 | 664 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h4| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198631.2 | NP_941033.2 | 4 - 5 | 664 | 73 | |
Set Membership: |
|