Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198631.2 | NP_941033.2 | 6 - 7 | 913 | 133 | |
| GenBank mRNA | AK129275.1 | - | 1 - 2 | 37 | 133 | |
| CF538051.1 | - | 2 - 3 | 127 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc3h4
Genomic Map