Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198631.2 | NP_941033.2 | 7 - 8 | 1058 | 62 | |
| GenBank mRNA | AK129275.1 | - | 2 - 3 | 182 | 62 | |
| CF538051.1 | - | 3 - 4 | 272 | 62 | ||
More Information
Set Membership: |
|

Zc3h4
Genomic Map