Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK129275.1 | - | 7 - 8 | 1252 | 73 | |
| BC060274.1 | - | 1 - 2 | 296 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h4| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK129275.1 | - | 7 - 8 | 1252 | 73 | |
| BC060274.1 | - | 1 - 2 | 296 | 73 | ||
Set Membership: |
|