Assay Details
| Gene Symbol: Slc1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009201.2 | NP_033227.2 | 3 - 4 | 1216 | 74 | |
| GenBank mRNA | AF246130.1 | - | 3 - 4 | 661 | 74 | |
| AK148346.1 | - | 3 - 4 | 1216 | 74 | ||
| AK157838.1 | - | 3 - 4 | 1195 | 74 | ||
| BC029873.1 | - | 3 - 4 | 719 | 4209657 | 74 | |
| BC037462.1 | - | 3 - 4 | 807 | 4192790 | 74 | |
| D85044.1 | - | 3 - 4 | 1149 | 74 | ||
| L42115.1 | - | 3 - 4 | 1192 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a5
Genomic Map