Assay Details
| Gene Symbol: Arhgap35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172739.4 | NP_766327.3 | 4 - 5 | 4616 | 101 | |
| GenBank mRNA | AK136766.1 | - | 3 - 4 | 1290 | 101 | |
| AK147326.1 | - | 4 - 5 | 4616 | 101 | ||
| AK147688.1 | - | 4 - 5 | 4616 | 101 | ||
| AK173242.1 | - | 3 - 4 | 3418 | 101 | ||
| BC035541.1 | - | 3 - 4 | 679 | 101 | ||
| BC139459.1 | - | 5 - 6 | 4394 | 8862481 | 101 | |
| BC139461.1 | - | 5 - 6 | 4253 | 8862483 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap35
Genomic Map