Assay Details
| Gene Symbol: Arhgap35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172739.4 | NP_766327.3 | 6 - 7 | 4849 | 99 | |
| GenBank mRNA | AK136766.1 | - | 5 - 6 | 1523 | 99 | |
| AK147326.1 | - | 6 - 7 | 4849 | 99 | ||
| AK147688.1 | - | 6 - 7 | 4849 | 99 | ||
| AK173242.1 | - | 5 - 6 | 3651 | 99 | ||
| BC035541.1 | - | 5 - 6 | 912 | 99 | ||
| BC139459.1 | - | 7 - 8 | 4627 | 8862481 | 99 | |
| BC139461.1 | - | 7 - 8 | 4486 | 8862483 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap35
Genomic Map