Assay Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110496.1 | NP_001103966.1 | 14 - 15 | 1366 | 82 | |
| NM_001110497.1 | NP_001103967.1 | 14 - 15 | 1366 | 82 | ||
| NM_173734.3 | NP_776095.2 | 14 - 15 | 1369 | 82 | ||
| GenBank mRNA | AK044591.1 | - | 14 - 15 | 1320 | 82 | |
| AK086738.1 | - | 14 - 15 | 1316 | 82 | ||
| AK088559.1 | - | 14 - 15 | 1351 | 82 | ||
| AK134268.1 | - | 14 - 15 | 1332 | 82 | ||
| BC004047.1 | - | 14 - 15 | 1315 | 82 | ||
| BC024676.1 | - | 4 - 5 | 317 | 82 | ||
| BC027354.1 | - | 14 - 15 | 1342 | 4010477 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem87a
Genomic Map