Assay Details
| Gene Symbol: Slc27a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011978.2 | NP_036108.2 | 5 - 6 | 1299 | 65 | |
| GenBank mRNA | AF033031.1 | - | 5 - 6 | 1180 | 65 | |
| AF072757.1 | - | 5 - 6 | 1177 | 65 | ||
| AJ223958.1 | - | 5 - 6 | 1217 | 65 | ||
| AK144069.1 | - | 5 - 6 | 1301 | 65 | ||
| AK144070.1 | - | 5 - 6 | 1213 | 65 | ||
| AK165446.1 | - | 5 - 6 | 1300 | 65 | ||
| BC013442.1 | - | 5 - 6 | 1282 | 4236616 | 65 | |
| BC022170.1 | - | 5 - 6 | 1278 | 4159081 | 65 | |
| BC024735.1 | - | 5 - 6 | 1268 | 5123650 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a2
Genomic Map