| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700017080.mm.1 |
NM_025392.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017080.mm.1 |
NM_025392.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017080.mm.1 |
TC0700001948.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017075.mm.1 |
NM_025392.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017075.mm.1 |
NM_025392.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017075.mm.1 |
TC0700001948.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017077.mm.1 |
NM_025392.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017077.mm.1 |
NM_025392.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017077.mm.1 |
TC0700001948.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017076.mm.1 |
NM_025392.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017076.mm.1 |
NM_025392.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017076.mm.1 |
TC0700001948.mm.1 |
MTA TC |