Assay Details
| Gene Symbol: Bccip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025392.2 | NP_079668.2 | 6 - 7 | 810 | 64 | |
| GenBank mRNA | AK003666.1 | - | 6 - 7 | 807 | 64 | |
| AK010684.1 | - | 6 - 7 | 804 | 64 | ||
| BC048465.1 | - | 6 - 7 | 802 | 6534043 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25542189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25542188 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140095 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100278 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132358 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158668 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170614 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148018 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542192 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170614 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158668 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132358 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148018 | - | - |
| Whole Mouse Genome G4122A | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | BC022920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK213507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK169005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK172504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443475 | AK135760.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120778 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158668 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170614 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132358 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06234493 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06234492 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06300520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25542188 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25542189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06415560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06404793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06622783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05905593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06415559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06528759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542190 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558327 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448542_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558325 | - | - |
| Mouse Genome 430 2.0 Array | 1448542_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448542_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66165_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66165_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700017086.mm.1 | NM_025392.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017086.mm.1 | NM_025392.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017086.mm.1 | TC0700001948.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017087.mm.1 | NM_025392.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017087.mm.1 | NM_025392.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017087.mm.1 | TC0700001948.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017088.mm.1 | NM_025392.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017088.mm.1 | NM_025392.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017088.mm.1 | TC0700001948.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448542_PM_at | - | - |
| Mouse Expression Set 430 | 1448542_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448542_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448542_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448542_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448542_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 863665 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106650112 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5270519 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2650750 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6660711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650750 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 670450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6660711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650750 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770003 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2759920 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2765373 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251737 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251737 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2770003 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255076 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2765373 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2759920 | - | - |
| mouse-6 v1.1 (Illumina) | scl000154.1_82-S | - | - |
| mouse-6 v1.1 (Illumina) | scl31959.7.1_44-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000028.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650750 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660711 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670450 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270519 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000028.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000154.1_82-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31959.7.1_44-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650750 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670450 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660711 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2759920 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770003 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2759920 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2862214 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2765373 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000028.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000154.1_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31959.7.1_44-S | - | - |

Bccip
Genomic Map