Assay Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024203.3 | NP_077165.2 | 2 - 3 | 1700 | 64 | |
| GenBank mRNA | AK016568.1 | - | 2 - 3 | 1666 | 64 | |
| AK132211.1 | - | 2 - 3 | 1678 | 64 | ||
| BC003200.1 | - | 1 - 2 | 603 | 64 | ||
| BC066088.1 | - | 2 - 3 | 1745 | 64 | ||
| DQ873694.1 | - | 2 - 3 | 1681 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120b
Genomic Map