Assay Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024203.3 | NP_077165.2 | 4 - 5 | 1981 | 73 | |
| NR_033586.1 | 4 - 5 | 3293 | 73 | |||
| GenBank mRNA | AK005957.1 | - | 1 - 2 | 87 | 73 | |
| AK016568.1 | - | 4 - 5 | 1947 | 73 | ||
| AK132211.1 | - | 4 - 5 | 1959 | 73 | ||
| AK187583.1 | - | 1 - 2 | 73 | |||
| AK220425.1 | - | 3 - 4 | 4568 | 73 | ||
| BC066088.1 | - | 4 - 5 | 2026 | 73 | ||
| BC085163.1 | - | 4 - 5 | 3247 | 30618952 | 73 | |
| DQ873694.1 | - | 4 - 5 | 1962 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005957.1 transcripts.
Set Membership: |
|

Fam120b
Genomic Map