Assay Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024203.3 | NP_077165.2 | 5 - 6 | 2160 | 68 | |
| NR_033586.1 | 5 - 6 | 3472 | 68 | |||
| GenBank mRNA | AK005957.1 | - | 2 - 3 | 266 | 68 | |
| AK016568.1 | - | 5 - 6 | 2126 | 68 | ||
| AK132211.1 | - | 5 - 6 | 2138 | 68 | ||
| AK187583.1 | - | 2 - 3 | 233 | 68 | ||
| BC003200.1 | - | 4 - 5 | 1063 | 68 | ||
| BC066088.1 | - | 5 - 6 | 2205 | 68 | ||
| BC085163.1 | - | 5 - 6 | 3426 | 30618952 | 68 | |
| DQ873694.1 | - | 5 - 6 | 2141 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120b
Genomic Map