Assay Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024203.3 | NP_077165.2 | 7 - 8 | 2458 | 64 | |
| NR_033586.1 | 7 - 8 | 3770 | 64 | |||
| GenBank mRNA | AK005957.1 | - | 4 - 5 | 564 | 64 | |
| AK016568.1 | - | 7 - 8 | 2424 | 64 | ||
| AK132211.1 | - | 7 - 8 | 2436 | 64 | ||
| AK180201.1 | - | 3 - 4 | 389 | 64 | ||
| BC003200.1 | - | 6 - 7 | 1361 | 64 | ||
| BC066088.1 | - | 7 - 8 | 2503 | 64 | ||
| BC085163.1 | - | 7 - 8 | 3724 | 30618952 | 64 | |
| DQ873694.1 | - | 7 - 8 | 2439 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120b
Genomic Map