Assay Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024203.3 | NP_077165.2 | 8 - 9 | 2563 | 62 | |
| NR_033586.1 | 8 - 9 | 3875 | 62 | |||
| GenBank mRNA | AK005957.1 | - | 5 - 6 | 669 | 62 | |
| AK016568.1 | - | 8 - 9 | 2529 | 62 | ||
| AK132211.1 | - | 8 - 9 | 2541 | 62 | ||
| BC066088.1 | - | 8 - 9 | 2608 | 62 | ||
| BC085163.1 | - | 8 - 9 | 3829 | 30618952 | 62 | |
| DQ873694.1 | - | 8 - 9 | 2544 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120b
Genomic Map