Assay Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024203.3 | NP_077165.2 | 9 - 10 | 2650 | 57 | |
| NR_033586.1 | 9 - 10 | 3962 | 57 | |||
| GenBank mRNA | AK005957.1 | - | 6 - 7 | 756 | 57 | |
| AK016568.1 | - | 9 - 10 | 2616 | 57 | ||
| AK132211.1 | - | 9 - 10 | 2628 | 57 | ||
| BC003200.1 | - | 8 - 9 | 1553 | 57 | ||
| BC066088.1 | - | 9 - 10 | 2695 | 57 | ||
| BC085163.1 | - | 9 - 10 | 3916 | 30618952 | 57 | |
| DQ873694.1 | - | 9 - 10 | 2631 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120b
Genomic Map