Assay Details
| Gene Symbol: Ifih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164477.1 | NP_001157949.1 | 9 - 10 | 2222 | 143 | |
| NM_027835.3 | NP_082111.2 | 10 - 11 | 2369 | 143 | ||
| GenBank mRNA | AF374384.1 | - | 10 - 11 | 2278 | 143 | |
| AK153018.1 | - | 10 - 11 | 2276 | 143 | ||
| AK153344.1 | - | 2 - 3 | 303 | 143 | ||
| AY075132.1 | - | 10 - 11 | 2044 | 143 | ||
| BC004031.1 | - | 3 - 4 | 511 | 143 | ||
| BC025508.1 | - | 5 - 6 | 813 | 143 | ||
| BC080200.1 | - | 9 - 10 | 2127 | 3597815 | 143 | |
| EF113547.1 | - | 10 - 11 | 2058 | 143 | ||
| EF113548.1 | - | 10 - 11 | 2058 | 143 | ||
| EU084750.1 | - | 10 - 11 | 2058 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifih1
Genomic Map