Assay Details
| Gene Symbol: Ifih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164477.1 | NP_001157949.1 | 10 - 11 | 2478 | 71 | |
| NM_027835.3 | NP_082111.2 | 11 - 12 | 2625 | 71 | ||
| GenBank mRNA | AF374384.1 | - | 11 - 12 | 2534 | 71 | |
| AK153018.1 | - | 11 - 12 | 2532 | 71 | ||
| AK153344.1 | - | 3 - 4 | 559 | 71 | ||
| AY075132.1 | - | 11 - 12 | 2300 | 71 | ||
| BC004031.1 | - | 4 - 5 | 767 | 71 | ||
| BC025508.1 | - | 6 - 7 | 1069 | 71 | ||
| BC080200.1 | - | 10 - 11 | 2383 | 3597815 | 71 | |
| EF113547.1 | - | 11 - 12 | 2314 | 71 | ||
| EF113548.1 | - | 11 - 12 | 2314 | 71 | ||
| EU084750.1 | - | 11 - 12 | 2314 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifih1
Genomic Map