| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700012119.mm.1 |
NM_001146010.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012119.mm.1 |
NM_001146010.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012119.mm.1 |
NM_199012.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012119.mm.1 |
NM_199012.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012119.mm.1 |
TC0700001423.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700012120.mm.1 |
NM_001146010.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012120.mm.1 |
NM_001146010.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012120.mm.1 |
NM_199012.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012120.mm.1 |
NM_199012.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012120.mm.1 |
TC0700001423.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700012117.mm.1 |
NM_001146010.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012117.mm.1 |
NM_001146010.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012117.mm.1 |
NM_199012.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012117.mm.1 |
NM_199012.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012117.mm.1 |
TC0700001423.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700012121.mm.1 |
NM_001146010.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012121.mm.1 |
NM_001146010.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012121.mm.1 |
NM_199012.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012121.mm.1 |
NM_199012.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012121.mm.1 |
TC0700001423.mm.1 |
MTA TC |