Assay Details
| Gene Symbol: Unc5cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152823.4 | NP_690036.4 | 6 - 7 | 1248 | 100 | |
| GenBank mRNA | AK144925.1 | - | 6 - 7 | 1249 | 100 | |
| AY510108.1 | - | 5 - 6 | 1115 | 100 | ||
More Information
Set Membership: |
|

Unc5cl
Genomic Map