Assay Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204071.1 | NP_001191000.1 | 4 - 5 | 308 | 138 | |
| NM_009420.2 | NP_033446.1 | 4 - 5 | 326 | 138 | ||
| GenBank mRNA | AK018856.1 | - | 4 - 5 | 257 | 138 | |
| BC049615.1 | - | 4 - 5 | 278 | 6742033 | 138 | |
| CJ046590.1 | - | 4 - 5 | 249 | 138 | ||
| M25533.1 | - | 4 - 5 | 278 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crisp2
Genomic Map