Assay Details
| Gene Symbol: Duoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042259.1 | - | 1 - 2 | 97 | ||
| BC019755.1 | - | 7 - 8 | 627 | 3964798 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Duoxa1
Genomic Map