Assay Details
| Gene Symbol: Adgre4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139138.3 | NP_631877.2 | 13 - 14 | 1667 | 150 | |
| GenBank mRNA | AF396935.1 | - | 13 - 14 | 1625 | 150 | |
| AK036459.1 | - | 13 - 14 | 1670 | 150 | ||
| AK037483.1 | - | 13 - 14 | 1612 | 150 | ||
| AK155935.1 | - | 13 - 14 | 1615 | 150 | ||
| AY032690.1 | - | 13 - 14 | 1586 | 150 | ||
| BC120538.1 | - | 13 - 14 | 1624 | 40129461 | 150 | |
| BC120540.1 | - | 13 - 14 | 1624 | 40129463 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgre4
Genomic Map