Assay Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301250.1 | NP_001288179.1 | 8 - 9 | 1266 | 68 | |
| NM_007602.4 | NP_031628.1 | 8 - 9 | 1430 | 68 | ||
| GenBank mRNA | AK164293.1 | - | 8 - 9 | 1340 | 68 | |
| AK170034.1 | - | 8 - 9 | 1367 | 68 | ||
| BC014767.1 | - | 8 - 9 | 1258 | 4458488 | 68 | |
| U85020.1 | - | 8 - 9 | 1349 | 68 | ||
| Y10656.1 | - | 8 - 9 | 1362 | 68 | ||
More Information
Set Membership: |
|

Capn5
Genomic Map