Assay Details
| Gene Symbol: Aars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198608.2 | NP_941010.2 | 6 - 7 | 1040 | 72 | |
| GenBank mRNA | AK146958.1 | - | 6 - 7 | 1040 | 72 | |
| AK167771.1 | - | 6 - 7 | 1035 | 72 | ||
| AK173139.1 | - | 2 - 3 | 306 | 72 | ||
| BC079844.1 | - | 6 - 7 | 1012 | 72 | ||
| BC113172.1 | - | 6 - 7 | 1036 | 40060953 | 72 | |
| BC113779.1 | - | 6 - 7 | 1036 | 40060952 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aars2
Genomic Map