Assay Details
| Gene Symbol: Tacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004468.2 | NP_001004468.1 | 7 - 8 | 6960 | 133 | |
| NM_021314.4 | NP_067289.2 | 3 - 4 | 1873 | 133 | ||
| NM_206856.3 | NP_996738.2 | 6 - 7 | 1839 | 133 | ||
| GenBank mRNA | AB041546.1 | - | 3 - 4 | 1874 | 133 | |
| AY177410.1 | - | 5 - 6 | 1851 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tacc2
Genomic Map