Assay Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159527.1 | NP_001152999.1 | 22 - 23 | 2809 | 101 | |
| NM_172470.3 | NP_766058.3 | 23 - 24 | 2842 | 101 | ||
| GenBank mRNA | AK083971.1 | - | 22 - 23 | 2754 | 101 | |
| BC056925.1 | - | 23 - 24 | 2951 | 5704780 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr35
Genomic Map