Assay Details
| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172671.2 | NP_766259.2 | 3 - 4 | 801 | 73 | |
| GenBank mRNA | AK044357.1 | - | 2 - 3 | 109 | 73 | |
| AY255619.1 | - | 2 - 3 | 141 | 73 | ||
| BE987673.1 | - | 3 - 4 | 338 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgr4
Genomic Map