Assay Details
| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172671.2 | NP_766259.2 | 7 - 8 | 1232 | 123 | |
| GenBank mRNA | AK044357.1 | - | 6 - 7 | 540 | 123 | |
| CF164628.1 | - | 3 - 4 | 254 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgr4
Genomic Map