Assay Details
| Gene Symbol: Rhob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148834.1 | - | 1 | 1200 | 151 | |
| AK164518.1 | - | 1 | 2148 | 151 | ||
| BC023334.1 | - | 1 | 1878 | 151 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rhob
Genomic Map