Assay Details
| Gene Symbol: Cstf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145529.3 | NP_663504.1 | 7 - 8 | 650 | 143 | |
| GenBank mRNA | AK159180.1 | - | 7 - 8 | 453 | 143 | |
| BC003241.1 | - | 7 - 8 | 611 | 3498424 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cstf3
Genomic Map