Assay Details
| Gene Symbol: Atp6v1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159632.1 | NP_001153104.1 | 8 - 9 | 750 | 64 | |
| NM_133699.2 | NP_598460.1 | 7 - 8 | 720 | 64 | ||
| GenBank mRNA | AB088357.1 | - | 7 - 8 | 705 | 64 | |
| AK004157.1 | - | 4 - 5 | 435 | 64 | ||
| BC056636.1 | - | 8 - 9 | 723 | 5065583 | 64 | |
| BC064071.1 | - | 7 - 8 | 689 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1c2
Genomic Map