Assay Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027959.3 | NP_082235.1 | 3 - 4 | 252 | 69 | |
| GenBank mRNA | AK005989.1 | - | 3 - 4 | 258 | 69 | |
| AK076558.1 | - | 3 - 4 | 253 | 69 | ||
| AK133792.1 | - | 3 - 4 | 259 | 69 | ||
| AK151008.1 | - | 3 - 4 | 233 | 69 | ||
| AK152960.1 | - | 3 - 4 | 265 | 69 | ||
| AK165877.1 | - | 3 - 4 | 231 | 69 | ||
| AK166111.1 | - | 3 - 4 | 259 | 69 | ||
| AK167386.1 | - | 3 - 4 | 270 | 69 | ||
| AK192703.1 | - | 2 - 3 | 161 | 69 | ||
| BC006865.1 | - | 3 - 4 | 241 | 2645183 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Pdia6
Genomic Map