Assay Details
| Gene Symbol: Myef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162417.1 | NP_001155889.1 | 17 - 18 | 1929 | 117 | |
| GenBank mRNA | AK048404.1 | - | 17 - 18 | 1896 | 117 | |
| AK143230.1 | - | 16 - 17 | 1814 | 164 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myef2
Genomic Map