Assay Details
| Gene Symbol: Muc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181729.2 | NP_859418.1 | 1404 | 63 | ||
| GenBank mRNA | BC120907.2 | - | 10 - 11 | 1292 | 40087870 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Muc6
Genomic Map