Assay Details
| Gene Symbol: Muc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181729.2 | NP_859418.1 | 3117 | 113 | ||
| GenBank mRNA | AJ010752.1 | - | 6 - 7 | 741 | 113 | |
| BC120907.2 | - | 23 - 24 | 3005 | 40087870 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Muc6
Genomic Map