Assay Details
| Gene Symbol: Muc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181729.2 | NP_859418.1 | 3653 | 68 | ||
| GenBank mRNA | AJ010752.1 | - | 9 - 10 | 1277 | 68 | |
| BC120907.2 | - | 26 - 27 | 3541 | 40087870 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Muc6
Genomic Map