Assay Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172281.2 | NP_758485.2 | 1 - 2 | 112 | 47 | |
| GenBank mRNA | AK129184.1 | - | 1 - 2 | 30 | 47 | |
| AK154982.1 | - | 1 - 2 | 43 | 47 | ||
| AK171462.1 | - | 1 - 2 | 41 | 47 | ||
| BC038348.1 | - | 1 - 2 | 26 | 4187484 | 47 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154982.1, AK171462.1, BC038348.1, NM_172281.2 transcripts.
Set Membership: |
|

Rnf40
Genomic Map