Assay Details
| Gene Symbol: Tead1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166584.1 | NP_001160056.1 | 3 - 4 | 138 | ||
| NM_001166585.1 | NP_001160057.1 | 2 - 3 | 321 | 75 | ||
| NM_009346.3 | NP_033372.1 | 3 - 4 | 728 | 75 | ||
| GenBank mRNA | AK140307.1 | - | 3 - 4 | 138 | ||
| AK148375.1 | - | 3 - 4 | 694 | 75 | ||
| BC052021.1 | - | 2 - 3 | 262 | 75 | ||
| BC060138.1 | - | 2 - 3 | 262 | 6410573 | 75 | |
| L06865.1 | - | 2 - 3 | 267 | 75 | ||
| L13853.1 | - | 3 - 4 | 455 | 75 | ||
| S74227.1 | - | 3 - 4 | 728 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tead1
Genomic Map